Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BMP15
Entrez Gene ID
448811
Gene Name
bone morphogenetic protein 15
Gene Aliases
GDF9B
Chromosome Location
Chr.X: 44613122 - 44620575 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005155.2 NP_001005155.2 2 - 2 1020 67
GenBank mRNA HQ450759.1 - 2 - 2 1020 67
HQ450760.1 - 2 - 2 773 67

View Full Product Details