Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IFN-DELTA-8
Entrez Gene ID
100153693
Gene Name
interferon-delta-8
Gene Aliases
-
Chromosome Location
Chr.1: 201277798 - 201278301 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164847.1 NP_001158319.1 1 - 1 50 88
GenBank mRNA GQ415081.1 - 1 - 1 50 88

View Full Product Details