The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001246282.1 | NP_001233211.1 | 1 - 2 | 293 | 94 |
GenBank mRNA | JF309103.1 | - | 1 - 2 | 293 | 94 |