Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1QTNF6
Entrez Gene ID
100217379
Gene Name
C1q and TNF related 6
Gene Aliases
-
Chromosome Location
Chr.5: 10601869 - 10606886 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142830.1 NP_001136302.1 2 - 3 335 82
GenBank mRNA AK234596.1 - 2 - 3 350 82
AK396426.1 - 2 - 3 347 82
FJ428537.1 - 2 - 3 335 82

View Full Product Details