Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LGALS2
Entrez Gene ID
100217393
Gene Name
galectin 2
Gene Aliases
Gal-2, Galectin-2
Chromosome Location
Chr.5: 10237772 - 10244555 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142842.1 NP_001136314.1 3 - 4 271 101
GenBank mRNA AK231447.1 - 3 - 4 303 101
FJ424704.1 - 3 - 4 271 101

View Full Product Details