Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLDN16
Entrez Gene ID
100302019
Gene Name
claudin 16
Gene Aliases
-
Chromosome Location
Chr.13: 127808599 - 127831081 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161644.2 NP_001155116.2 1 - 2 350 59
XM_021068343.1 XP_020924002.1 - 314 59
GenBank mRNA FJ873105.1 - 1 - 2 350 59

View Full Product Details