Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG9B
Entrez Gene ID
100462720
Gene Name
autophagy related 9B
Gene Aliases
-
Chromosome Location
Chr.18: 6200246 - 6208988 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190280.1 NP_001177209.1 9 - 10 2298 95
XM_021078467.1 XP_020934126.1 - 2687 95
XM_021078468.1 XP_020934127.1 - 2511 95
GenBank mRNA HM046519.1 - 9 - 10 2298 95

View Full Product Details