Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ODF4
Entrez Gene ID
100529212
Gene Name
outer dense fiber of sperm tails 4
Gene Aliases
-
Chromosome Location
Chr.12: 53517175 - 53525458 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202461.1 NP_001189390.1 2 - 3 122 108
GenBank mRNA HQ834533.1 - 2 - 3 122 108

View Full Product Details