Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDCBP2
Entrez Gene ID
100513633
Gene Name
syndecan binding protein 2
Gene Aliases
-
Chromosome Location
Chr.17: 33967075 - 33985975 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244863.1 NP_001231792.1 8 - 9 975 90
XM_013985196.2 XP_013840650.1 - 1307 90
XM_021077266.1 XP_020932925.1 - 1972 90
XM_021077267.1 XP_020932926.1 - 1252 90
GenBank mRNA AK349737.1 - 8 - 9 977 90

View Full Product Details