Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MPC1
Entrez Gene ID
100513634
Gene Name
mitochondrial pyruvate carrier 1
Gene Aliases
BRP44L
Chromosome Location
Chr.1: 2541475 - 2552864 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244864.1 NP_001231793.1 4 - 5 399 68
XM_013984353.2 XP_013839807.1 - 439 68
XM_021071721.1 XP_020927380.1 - 2358 68
GenBank mRNA AK233047.1 - 4 - 5 399 68
AK391807.1 - 4 - 5 431 68
AK396434.1 - 4 - 5 366 68
AK399856.1 - 4 - 5 444 68

View Full Product Details