Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAYN
Entrez Gene ID
100513941
Gene Name
layilin
Gene Aliases
-
Chromosome Location
Chr.9: 39306276 - 39326825 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244894.1 NP_001231823.1 2 - 3 551 82
GenBank mRNA AK240128.1 - 1 - 2 250 82
FD642694.1 - 2 - 3 556 82

View Full Product Details