Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPY30
Entrez Gene ID
100514043
Gene Name
dpy-30 histone methyltransferase complex regulatory subunit
Gene Aliases
-
Chromosome Location
Chr.3: 107520641 - 107533504 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244925.1 NP_001231854.1 3 - 4 175 129
XM_005655249.3 XP_005655306.1 - 314 129
GenBank mRNA AK346618.1 - 3 - 4 175 129
AK392665.1 - 3 - 4 176 129

View Full Product Details