Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBX2
Entrez Gene ID
100622658
Gene Name
T-box transcription factor 2
Gene Aliases
-
Chromosome Location
Chr.12: 36878662 - 36887513 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246248.1 NP_001233177.1 1 - 2 414 68
GenBank mRNA AK397678.1 - 1 - 2 114 68
HQ630069.1 - 1 - 2 414 68

View Full Product Details