Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOX
Entrez Gene ID
100525278
Gene Name
lysyl oxidase
Gene Aliases
-
Chromosome Location
Chr.2: 125595115 - 125607026 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206403.1 NP_001193332.1 3 - 4 899 74
GenBank mRNA AK395814.1 - 3 - 4 899 74
CK452593.1 - 4 - 5 683 74

View Full Product Details