Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
TADA3
Entrez Gene ID
100157464
Gene Name
transcriptional adaptor 3
Gene Aliases
-
Chromosome Location
Chr.13: 66067623 - 66080170 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206536.1 NP_001193465.1 6 - 7 998 63
GenBank mRNA AK233113.1 - 6 - 7 957 63
AK235953.1 - 6 - 7 998 63
AK238770.1 - 6 - 7 903 63

View Full Product Details