Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADGRG1
Entrez Gene ID
100515029
Gene Name
adhesion G protein-coupled receptor G1
Gene Aliases
GPR56
Chromosome Location
Chr.6: 19523618 - 19570891 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243056.1 NP_001229985.1 8 - 9 1211 86
XM_021093328.1 XP_020948987.1 - 1318 86
XM_021093330.1 XP_020948989.1 - 1417 86
XM_021093331.1 XP_020948990.1 - 1163 86
XM_021093332.1 XP_020948991.1 - 1139 86
XM_021093333.1 XP_020948992.1 - 1613 86
XM_021093334.1 XP_020948993.1 - 1335 86
XM_021093335.1 XP_020948994.1 - 1193 86
XM_021093336.1 XP_020948995.1 - 2749 86
XM_021093337.1 XP_020948996.1 - 1372 86
XM_021093338.1 XP_020948997.1 - 1287 86
GenBank mRNA AK348726.1 - 8 - 9 1211 86
AK349595.1 - 8 - 9 1430 86

View Full Product Details