Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP1S
Entrez Gene ID
100517999
Gene Name
microtubule associated protein 1S
Gene Aliases
-
Chromosome Location
Chr.2: 60003116 - 60043133 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243185.1 NP_001230114.1 3 - 4 348 76
XM_021079065.1 XP_020934724.1 - 932 76
GenBank mRNA AK235554.1 - 3 - 4 326 76

View Full Product Details