Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MMGT1
Entrez Gene ID
100520118
Gene Name
membrane magnesium transporter 1
Gene Aliases
-
Chromosome Location
Chr.X: 111145712 - 111158298 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243208.1 NP_001230137.1 3 - 4 300 96
GenBank mRNA AK344491.1 - 3 - 4 269 96
FS668658.1 - 3 - 4 301 96

View Full Product Details