Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPIFA3
Entrez Gene ID
100155470
Gene Name
BPI fold containing family A member 3
Gene Aliases
-
Chromosome Location
Chr.17: 36738532 - 36749816 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243255.1 NP_001230184.1 5 - 6 803 68
XM_021077227.1 XP_020932886.1 - 756 68
GenBank mRNA ES446305.1 - 5 - 6 631 68

View Full Product Details