Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARV1
Entrez Gene ID
100151849
Gene Name
ARV1 homolog, fatty acid homeostasis modulator
Gene Aliases
-
Chromosome Location
Chr.14: 59396958 - 59409008 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243293.1 NP_001230222.1 1 - 2 233 114
GenBank mRNA AK348219.1 - 1 - 2 233 114
AK391940.1 - 1 - 2 218 114
AY610027.1 - 1 - 2 178 114
FS700419.1 - 1 - 2 198 114

View Full Product Details