Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL13
Entrez Gene ID
100628232
Gene Name
ribosomal protein L13
Gene Aliases
-
Chromosome Location
Chr.6: 410923 - 413396 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243345.1 NP_001230274.1 2 - 3 182 150
GenBank mRNA AK231738.1 - 1 - 2 340 150
AK237333.1 - 2 - 3 155 150
AK347687.1 - 2 - 3 148 150
AK350211.1 - 1 - 2 335 150
BP171914.1 - 2 - 3 185 150

View Full Product Details