Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LACTB
Entrez Gene ID
100152773
Gene Name
lactamase beta
Gene Aliases
-
Chromosome Location
Chr.1: 108909690 - 108924509 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243362.1 NP_001230291.1 1 - 2 391 90
XM_021086851.1 XP_020942510.1 - 415 90
GenBank mRNA AK351983.1 - 1 - 2 397 90

View Full Product Details