Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLDN10
Entrez Gene ID
100153752
Gene Name
claudin 10
Gene Aliases
-
Chromosome Location
Chr.11: 65027706 - 65145002 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243444.1 NP_001230373.1 1 - 2 229 61
GenBank mRNA AK352098.1 - 1 - 2 229 61
AK391886.1 - 1 - 2 249 61
AK399963.1 - 1 - 2 313 61
AY609541.1 - 1 - 2 292 61

View Full Product Details