Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCEH1
Entrez Gene ID
100154209
Gene Name
neutral cholesterol ester hydrolase 1
Gene Aliases
-
Chromosome Location
Chr.13: 111148677 - 111207285 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243484.1 NP_001230413.1 3 - 4 556 89
GenBank mRNA AK344477.1 - 3 - 4 556 89

View Full Product Details