Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INTS13
Entrez Gene ID
100155867
Gene Name
integrator complex subunit 13
Gene Aliases
ASUN, C5H12orf11
Chromosome Location
Chr.5: 46823746 - 46860552 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243595.1 NP_001230524.1 2 - 3 431 102
XM_021090951.1 XP_020946610.1 - 765 102
XM_021090952.1 XP_020946611.1 - 376 102
XM_021090953.1 XP_020946612.1 - 356 102
XM_021090954.1 XP_020946613.1 - 380 102
GenBank mRNA AK343638.1 - 2 - 3 431 102

View Full Product Details