Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDUFAF3
Entrez Gene ID
100156018
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 3
Gene Aliases
-
Chromosome Location
Chr.13: 31656941 - 31658087 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243606.1 NP_001230535.1 4 - 5 463 99
GenBank mRNA AK350073.1 - 4 - 5 449 99
EW001058.2 - 4 - 5 470 99
EW089413.2 - 1 - 2 63 99

View Full Product Details