Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLN6
Entrez Gene ID
100156072
Gene Name
CLN6 transmembrane ER protein
Gene Aliases
-
Chromosome Location
Chr.1: 166107895 - 166126054 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243614.1 NP_001230543.1 6 - 7 778 76
XM_021095640.1 XP_020951299.1 - 687 76
GenBank mRNA AK233319.1 - 6 - 7 778 76

View Full Product Details