Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSD17B13
Entrez Gene ID
100626886
Gene Name
hydroxysteroid 17-beta dehydrogenase 13
Gene Aliases
-
Chromosome Location
Chr.8: 131555517 - 131570067 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243622.1 NP_001230551.1 4 - 5 585 96
XM_021100397.1 XP_020956056.1 - 733 96
XM_021100398.1 XP_020956057.1 - 741 96
GenBank mRNA AK232809.1 - 4 - 5 585 96

View Full Product Details