Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C5H12orf29
Entrez Gene ID
100157029
Gene Name
chromosome 5 C12orf29 homolog
Gene Aliases
-
Chromosome Location
Chr.5: 94473172 - 94489643 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243635.1 NP_001230564.1 6 - 7 868 99
XM_013988455.2 XP_013843909.1 - 866 99
XM_021090957.1 XP_020946616.1 - 1085 99
GenBank mRNA AK343312.1 - 6 - 7 868 99

View Full Product Details