Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM216B
Entrez Gene ID
100157364
Gene Name
family with sequence similarity 216 member B
Gene Aliases
C11H13orf30
Chromosome Location
Chr.11: 24312382 - 24321943 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243662.1 NP_001230591.1 4 - 5 387 106
XM_005656846.3 XP_005656903.1 - 344 106
XM_005656847.3 XP_005656904.1 - 344 106
XM_013989774.2 XP_013845228.1 - 473 106
XM_021065104.1 XP_020920763.1 - 392 106
XM_021065105.1 XP_020920764.1 - 533 106
XM_021065106.1 XP_020920765.1 - 527 106
XM_021065107.1 XP_020920766.1 - 545 106
XM_021065109.1 XP_020920768.1 - 373 106
GenBank mRNA AK347614.1 - 4 - 5 362 106
BW967519.1 - 4 - 5 474 106

View Full Product Details