Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMED5
Entrez Gene ID
100157999
Gene Name
transmembrane p24 trafficking protein 5
Gene Aliases
-
Chromosome Location
Chr.4: 124009825 - 124027445 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243695.1 NP_001230624.1 1 - 2 340 88
GenBank mRNA AY610000.1 - 1 - 2 340 88

View Full Product Details