The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243695.1 | NP_001230624.1 | 3 - 4 | 625 | 97 |
GenBank mRNA | AY610000.1 | - | 3 - 4 | 625 | 97 |