Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPR82
Entrez Gene ID
100158124
Gene Name
G protein-coupled receptor 82
Gene Aliases
-
Chromosome Location
Chr.X: 37328672 - 37334144 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243706.1 NP_001230635.1 2 - 3 320 139
GenBank mRNA AK230470.1 - 2 - 3 319 139
AK389657.1 - 2 - 3 323 139
BW956594.1 - 2 - 3 323 139

View Full Product Details