Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXOSC7
Entrez Gene ID
100511347
Gene Name
exosome component 7
Gene Aliases
-
Chromosome Location
Chr.13: 28176806 - 28206435 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243824.1 NP_001230753.1 2 - 3 167 77
GenBank mRNA AK395369.1 - 2 - 3 186 77
AY610143.1 - 2 - 3 167 77

View Full Product Details