Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRACR2B
Entrez Gene ID
100511435
Gene Name
calcium release activated channel regulator 2B
Gene Aliases
EFCAB4A
Chromosome Location
Chr.2: 518102 - 524085 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243838.1 NP_001230767.1 3 - 4 370 105
XM_013989677.2 XP_013845131.1 - 1048 105
XM_013989679.2 XP_013845133.1 - 1012 105
XM_013989682.2 XP_013845136.1 - 1048 105
XM_013989684.2 XP_013845138.1 - 1140 105
GenBank mRNA AK350005.1 - 3 - 4 370 105

View Full Product Details