Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASPDH
Entrez Gene ID
100511638
Gene Name
aspartate dehydrogenase domain containing
Gene Aliases
-
Chromosome Location
Chr.6: 55341343 - 55343537 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243870.1 NP_001230799.1 5 - 6 723 70
GenBank mRNA CF175854.1 - 2 - 3 335 70

View Full Product Details