Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CAMK1G
Entrez Gene ID
100512553
Gene Name
calcium/calmodulin dependent protein kinase IG
Gene Aliases
-
Chromosome Location
Chr.9: 133351523 - 133380454 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243934.1 NP_001230863.1 6 - 7 666 101
XM_021102014.1 XP_020957673.1 - 1621 101
XM_021102015.1 XP_020957674.1 - 1824 101
XM_021102016.1 XP_020957675.1 - 1546 101
XM_021102017.1 XP_020957676.1 - 1476 101
XM_021102018.1 XP_020957677.1 - 1621 101
XM_021102019.1 XP_020957678.1 - 1476 101
GenBank mRNA AK349243.1 - 7 - 8 723 101

View Full Product Details