Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GNPDA1
Entrez Gene ID
100512094
Gene Name
glucosamine-6-phosphate deaminase 1
Gene Aliases
-
Chromosome Location
Chr.2: 143604362 - 143616037 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244093.1 NP_001231022.1 6 - 7 820 68
XM_013995256.2 XP_013850710.1 - 1351 68
GenBank mRNA AK349993.1 - 6 - 7 811 68

View Full Product Details