Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CXCL14
Entrez Gene ID
494467
Gene Name
C-X-C motif chemokine ligand 14
Gene Aliases
BRAK
Chromosome Location
Chr.2: 137803988 - 137813118 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244128.1 NP_001231057.1 2 - 3 417 90
GenBank mRNA AK231165.1 - 2 - 3 455 90
AK231389.1 - 2 - 3 447 90
AK231559.1 - 2 - 3 417 90
AK233754.1 - 2 - 3 379 90
AK233879.1 - 2 - 3 382 90
AK234044.1 - 2 - 3 385 90
AK345106.1 - 2 - 3 379 90
AK345135.1 - 2 - 3 382 90
AK345275.1 - 2 - 3 382 90
AK348211.1 - 2 - 3 380 90

View Full Product Details