Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SSR3
Entrez Gene ID
100625983
Gene Name
signal sequence receptor subunit 3
Gene Aliases
-
Chromosome Location
Chr.13: 96349782 - 96390190 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244147.1 NP_001231076.1 3 - 4 456 118
GenBank mRNA AK231884.1 - 3 - 4 458 118
AK232862.1 - 3 - 4 456 118
AK345822.1 - 3 - 4 422 118
AK350259.1 - 3 - 4 421 118
AY610158.1 - 3 - 4 430 118

View Full Product Details