Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFEMP1
Entrez Gene ID
100512046
Gene Name
EGF containing fibulin extracellular matrix protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 85407677 - 85473251 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244261.1 NP_001231190.1 1 - 2 121 103
XM_013996146.2 XP_013851600.1 - 117 103
GenBank mRNA DT331816.1 - 1 - 2 159 103
FD638968.1 - 1 - 2 79 103
HQ630067.1 - 1 - 2 141 103

View Full Product Details