Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PGLYRP4
- Entrez Gene ID
-
100689249
- Gene Name
- peptidoglycan recognition protein 4
- Gene Aliases
-
-
- Chromosome Location
-
-
- Gene Symbol
- PGLYRP4
- Entrez Gene ID
-
396556
- Gene Name
- peptidoglycan recognition protein 4
- Gene Aliases
-
PPGRP-S
- Chromosome Location
-
Chr.4: 96243174 - 96247517 on Build Sscrofa11.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB292178.1 |
- |
5 - 6 |
658 |
75 |
View Full Product Details