Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B4GALNT2
Entrez Gene ID
100621328
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.12: 25368234 - 25410021 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244330.1 NP_001231259.1 10 - 11 1356 97
GenBank mRNA AK236764.1 - 10 - 11 1356 97
AK397757.1 - 11 - 12 1432 97
KF501048.1 - 10 - 11 1358 97
KU358546.1 - 9 - 10 1304 97

View Full Product Details