Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTP4A2
Entrez Gene ID
100625391
Gene Name
protein tyrosine phosphatase 4A2
Gene Aliases
-
Chromosome Location
Chr.6: 88419444 - 88452757 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244341.1 NP_001231270.1 4 - 5 441 100
XM_021093389.1 XP_020949048.1 - 1398 100
XM_021093390.1 XP_020949049.1 - 1090 100
GenBank mRNA AK343562.1 - 5 - 6 1391 100
AK393251.1 - - 592 100
AK395502.1 - 5 - 6 1379 100
FD613790.1 - 4 - 5 441 100

View Full Product Details