Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPRG1L
Entrez Gene ID
100512473
Gene Name
tumor protein p63 regulated 1 like
Gene Aliases
-
Chromosome Location
Chr.6: 65150897 - 65154311 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244355.1 NP_001231284.1 2 - 3 389 71
NM_001244356.1 NP_001231285.1 2 - 3 245 71
GenBank mRNA AK239234.1 - 2 - 3 376 71
AK391650.1 - 2 - 3 384 71
DN115724.1 - 2 - 3 245 71
FD595047.1 - 2 - 3 217 71
FS646464.1 - 2 - 3 395 71
FS665120.1 - 2 - 3 384 71

View Full Product Details