Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
SLC35D2
Entrez Gene ID
100514312
Gene Name
solute carrier family 35 member D2
Gene Aliases
-
Chromosome Location
Chr.10: 25738208 - 25789487 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244362.1 NP_001231291.1 8 - 9 709 103
XM_021063996.1 XP_020919655.1 - 899 103
XM_021063997.1 XP_020919656.1 - 827 103
GenBank mRNA AK238334.1 - 8 - 9 709 103

View Full Product Details