Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPD52
Entrez Gene ID
100151928
Gene Name
tumor protein D52
Gene Aliases
-
Chromosome Location
Chr.4: 56472992 - 56603616 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244387.1 NP_001231316.1 2 - 3 197 116
XM_021088495.1 XP_020944154.1 - 642 116
XM_021088496.1 XP_020944155.1 - 629 116
XM_021088497.1 XP_020944156.1 - 583 116
XM_021088499.1 XP_020944158.1 - 163 116
XM_021088500.1 XP_020944159.1 - 318 116
XM_021088501.1 XP_020944160.1 - 317 116
XM_021088502.1 XP_020944161.1 - 579 116
GenBank mRNA AK237145.1 - 2 - 3 188 116
AK396598.1 - 2 - 3 220 116
AY610035.1 - 2 - 3 197 116

View Full Product Details