Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NFATC2IP
Entrez Gene ID
100625065
Gene Name
nuclear factor of activated T cells 2 interacting protein
Gene Aliases
-
Chromosome Location
Chr.3: 18609943 - 18624150 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244522.1 NP_001231451.1 2 - 3 468 69
XM_005662047.3 XP_005662104.2 - 553 69
GenBank mRNA AK235025.1 - 2 - 3 468 69
AK395742.1 - 2 - 3 472 69

View Full Product Details