Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRDX3
Entrez Gene ID
100154143
Gene Name
peroxiredoxin 3
Gene Aliases
-
Chromosome Location
Chr.14: 129079050 - 129090790 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244531.1 NP_001231460.1 4 - 5 476 102
GenBank mRNA AK351883.1 - 4 - 5 476 102
AK392207.1 - 4 - 5 506 102
AK392777.1 - 4 - 5 486 102
AK398410.1 - 3 - 4 218 102

View Full Product Details