Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CMTM5
Entrez Gene ID
100514659
Gene Name
CKLF like MARVEL transmembrane domain containing 5
Gene Aliases
-
Chromosome Location
Chr.7: 75707132 - 75709809 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244546.1 NP_001231475.1 2 - 3 500 60
GenBank mRNA AY609760.1 - 2 - 3 485 60
BE235279.1 - 2 - 3 500 60
EW187622.2 - 1 - 2 129 60

View Full Product Details